missing translation for 'onlineSavingsMsg'
Learn More

Dipraglurant, MedChemExpress

Product Code. p-200098158
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481449

Brand: Medchem Express HY14859

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dipraglurant (ADX48621) is a potent, selective, orally active and brain penetrant mGluR5 negative allosteric modulator (NAM), with an IC50 of 21 nM. Dipraglurant can reduce Levodopa-induced dyskinesia (LID) in vivo.

Dipraglurant (ADX48621) is a potent, selective, orally active and brain penetrant mGluR5 negative allosteric modulator (NAM), with an IC50 of 21 nM. Dipraglurant can reduce Levodopa-induced dyskinesia (LID) in vivo.

Specifications

Chemical Name or Material Dipraglurant
CAS 872363-17-2
Color White
Purity Grade Notes Research
Molecular Formula C16H12FN3
Quantity 1 mg
Synonym ADX48621
Solubility Information DMSO : ≥ 40 mg/mL (150.78 mM)
SMILES FC1=CN2C(C=C1)=NC(CCC#CC3=NC=CC=C3)=C2
Molecular Weight (g/mol) 265.29
Formula Weight 265.29
Percent Purity 95.95%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.