Learn More
Diphenylpyraline, TRC
Product Code. 30380526
Change view
Click to view available options
Quantity:
50 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380526 50 mg
30374290 500 mg
30374532 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30380526 Supplier Toronto Research Chemicals Supplier No. TRCD49211050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 147-20-6
Molecular Formula C19 H23 N O
Molecular Weight (g/mol) 281.39
Synonym Diphenylpyraline, 4-(Diphenylmethoxy)-1-methylpiperidine, 1-Methyl-4-piperidyl benzhydryl ether, 4-(Benzhydryloxy)-1-methylpiperidine, AN 1041, Diphenpyraline, Diphenylpyraline, Diphenylpyrilene, Hystryl, Lyssipoll, Mepiben, N-Methyl-4-(benzhydryloxy)piperidine, Neargal, P 253
IUPAC Name 4-benzhydryloxy-1-methylpiperidine
SMILES CN1CCC(CC1)OC(c2ccccc2)c3ccccc3
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C19H23NO/c1-20-14-12-18(13-15-20)21-19(16-8-4-2-5-9-16)17-10-6-3-7-11-17/h2-11,18-19H,12-15H2,1H3
Formula Weight 281.178
Percent Purity >95%
Chemical Name or Material Diphenylpyraline
Grade HPLC