Learn More
Diphenylphosphine, 95%, Thermo Scientific Chemicals
Product Code. 10206693 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10206693 5 g 5g
10023993 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10206693 Supplier Thermo Scientific Acros Supplier No. 320970050
 Restricted Item
In Stock
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 829-85-6
Molecular Weight (g/mol) 186.19
MDL Number MFCD00003040
InChI Key GPAYUJZHTULNBE-UHFFFAOYSA-N
IUPAC Name diphenylphosphane
SMILES P(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Density 1.0700 g/mL
Boiling Point 280.0°C
Flash Point 110°C
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C12H11P
Linear Formula C6H5PHC6H5
Refractive Index 1.6240 to 1.6260
Quantity 5 g
Merck Index 15, 3364
Specific Gravity 1.07
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 186.19
Chemical Name or Material Diphenylphosphine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diphenylphosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Pyrophoric liquid Category 1
Hazard Statement
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H250-Catches fire spontaneously if exposed to air.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.