Learn More
Dioctyltin oxide, Sigma-Aldrich™
Product Code. 32238313
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238313 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238313 Supplier Sigma Aldrich Supplier No. 8035440025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dioctyltin oxide
CAS 870-08-6
Boiling Point 230°C (1013 hPa)
Flash Point 70°C
Quantity 25 g
Synonym Dioctyltin oxide, Dioctyloxostannane, Dioctyltin oxide
Molecular Weight (g/mol) 361.15
Grade Not Available