Learn More
DIN EN 17892:2024-08 PFAS Mixture 10 μg/mL in Methanol:Water, Dr. Ehrenstorfer
Product Code. 30312526
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312526 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30312526 Supplier Dr. Ehrenstorfer Supplier No. DREA30000080MW
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 39108-34-4, 757124-72-4, 27619-97-2, 2991-50-6, 919005-14-4, 756426-58-1
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol/Water