Learn More
Lenz Laborglasintrumente™ Dimroth Condensers with Glass Olives
Product Code. 10090271
Change view
Click to view available options
Length (Metric) Jacket:
250 mm
400 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Jacket unitSize
10090271 250 mm Each
10400713 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10090271 Supplier Lenz Laborglasintrumente™ Supplier No. 05240306
  • ONLINE EXCLUSIVE PRICE
    1725.00 NOK
    1696.73 NOK / Pack of 1
    Save 28.27 NOK
    1% Off
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Dimroth condensers for condensing vapours
  • Dimroth condensers acc. to DIN 12591 and in addition to DIN standard
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Description Cooling Surface - 810 cm²
For Use With (Application) Condensing vapours
Length (Metric) Jacket 250 mm
Quantity 1
Standard Taper Joint No NS 45/40
Material DURAN™ Tubing