Learn More
Lenz Laborglasintrumente™ Dimroth Condensers For Extractors
Product Code. 10149560
Change view
Click to view available options
Standard Taper Joint No:
NS 60/46
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
10149560 NS 60/46 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10149560 Supplier Lenz Laborglasintrumente™ Supplier No. 05246007
  • ONLINE EXCLUSIVE PRICE
    2540.00 NOK
    2103.81 NOK / Pack of 1
    Save 436.19 NOK
    17% Off
Estimated Shipment: 05-01-2027
Dimroth condensers for extractors, for condensing vapours
  • With NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Description Cooling Surface - 680 cm²
For Use With (Application) Condensing vapours
For Use With (Equipment) Extractors 300 - 500 mL
Quantity 1
Standard Taper Joint No NS 60/46
Material DURAN™ Tubing