Learn More
Lenz Laborglasintrumente Dimroth Condensers for Extractors, with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05246005
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 10393191
  • 1197.80 NOK / Each
Estimated Shipment: 28-06-2024
Description
  • Dimroth condensers for extractors
  • For condensing vapours
  • With NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Specifications
Glass olives
34/35 NS
1 Ea.
Duran Glass