Learn More
Dimethylformamide - D7, deuteration degree min. 99.5% for NMR spectroscopy MagniSolv™, MilliporeSigma™
Product Code. 15805600
Change view
Click to view available options
Quantity:
10 x 0.75 mL
Packaging:
Glass Ampule
Unit Size:
7.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15805600 10 x 0.75 mL Glass Ampule 7.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15805600 Supplier Sigma Aldrich Supplier No. 1116560009
 Restricted Item
Estimated Shipment: 26-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
For NMR Spectroscopy
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4472-41-7
Molecular Formula C3H7NO
Molecular Weight (g/mol) 80.138
InChI Key ZMXDDKWLCZADIW-YYWVXINBSA-N
Synonym n,n-dimethylformamide-d7, dmf-d7, deuterated dmf, n,n-di 2h3 methyl 2h formamide, dimethyl formamide-d7, dimethylformamide d, heptadeutero-n,n-dimethylformamide, n,n-dimethylformamide-d7 dmf-d7, formamide-1-d, n,n-di methyl-d3, n,n-dimethylformamide-d7 99.5atom%d
PubChem CID 78225
IUPAC Name 1-deuterio-N,N-bis(trideuteriomethyl)formamide
SMILES CN(C)C=O
Specifications
Physical Form Liquid
Color Colorless
Odor Weakly Amine-like
Packaging Glass Ampule
Vapor Pressure 3.77 hPa at 20°C (68°F)
Quantity 10 x 0.75 mL
Assay Percent Range 99.5% min.
Formula Weight 80.14
Chemical Name or Material Dimethylformamide - D7