missing translation for 'onlineSavingsMsg'
Learn More

Dimethylaminoacetaldehyde diethyl acetal, 95%, Thermo Scientific Chemicals

Product Code. p-7044408
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302446 5 g 5g
11312446 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302446 Supplier Thermo Scientific Alfa Aesar Supplier No. L06272.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3616-56-6
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.245
MDL Number MFCD00009232
InChI Key SSFAUOAQOOISRQ-UHFFFAOYSA-N
Synonym dimethylamino acetaldehyde diethyl acetal, 2,2-diethoxy-n,n-dimethylethylamine, 2,2-diethoxyethyl dimethylamine, dimethylaminoacetaldehyde diethyl acetal, ethanamine, 2,2-diethoxy-n,n-dimethyl, dimethylaminoacetaldehyde diethylacetal, n-2,2-diethoxyethyl dimethylamine, 2,2-dieth, acmc-1ajh8
PubChem CID 77163
IUPAC Name 2,2-diethoxy-N,N-dimethylethanamine
SMILES CCOC(CN(C)C)OCC

Specifications

Density 0.876
Boiling Point 164°C to 166°C
Flash Point 45°C (113°F)
Odor Amine-like
Refractive Index 1.4115
Quantity 5 g
UN Number UN1993
Beilstein 1739149
Formula Weight 161.25
Percent Purity 95%
Chemical Name or Material Dimethylaminoacetaldehyde diethyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.