missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Phthalate, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7072993 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10364980

Brand: Fisher Chemical D/4050/08

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 131-11-3
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008425
InChI Key NIQCNGHVCWTJSM-UHFFFAOYSA-N
Synonym dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm
PubChem CID 8554
ChEBI CHEBI:4609
IUPAC Name dimethyl benzene-1,2-dicarboxylate
SMILES COC(=O)C1=CC=CC=C1C(=O)OC

Specifications

Melting Point 2°C
Color Colorless
Boiling Point 282°C
Packaging Amber glass bottle
Quantity 500 mL
Vapor Density 6.69
Formula Weight 194.19g/mol
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.