Learn More
Dimethyl adipate, 99%
Product Code. p-7025599
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
Product Code. 11344489
Brand: Thermo Scientific Alfa Aesar B21174.18
Request Bulk or Custom Format
CAS: 627-93-0 | C8H14O4 | 174.196 g/mol
Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl adipate is used in cosmetics (in emollients and skin conditioning). It acts as a cosmetic plasticizer. It is also used in agrochemicals and dye as well as a precursor for the preparation of active pharmaceutical ingredients. Further, it serves as a polymer intermediate. In addition to this, it is employed as a solvent for paint stripping and resins.
Solubility
Miscible with alcohols and ether. Immiscible with water.
Notes
Moisture sensitive. Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 627-93-0
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00008469
InChI Key UDSFAEKRVUSQDD-UHFFFAOYSA-N
Synonym dimethyl adipate, dimethyladipate, adipic acid dimethyl ester, hexanedioic acid, dimethyl ester, adipic acid, dimethyl ester, 1,6-dimethylhexanedioate, methyl adipate van, 1,6-dimethyl hexanedioate, dbe 6, unii-by71rx0r62
PubChem CID 12329
ChEBI CHEBI:34715
IUPAC Name dimethyl hexanedioate
SMILES COC(=O)CCCCC(=O)OC
Specifications
Melting Point 8°C
Density 1.063
Boiling Point 109°C to 110°C (14 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.428
Quantity 50 g
Beilstein 1707443
Merck Index 14,162
Solubility Information Miscible with alcohols and ether. Immiscible with water.
Formula Weight 174.2
Percent Purity 99%
Chemical Name or Material Dimethyl adipate