Learn More
Dimethomorph, TRC
Product Code. 30041345
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041345 1 g 1g
30005694 10 g 10g
18032630 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30041345 Supplier Toronto Research Chemicals Supplier No. TRCD4605301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110488-70-5
Molecular Formula C21 H22 Cl N O4
Molecular Weight (g/mol) 387.86
Synonym Morpholine, 4-[3-(4-chlorophenyl)-3-(3,4-dimethoxyphenyl)-1-oxo-2-propenyl]- (9CI), 3-(4-Chlorophenyl)-3-(3,4-dimethoxyphenyl)-1-(4-morpholinyl)-2-propen-1-one, Acrobat, Akrobat, Athlete, CME 151, Dimethomorph, Festival C, Forum, Forum (biocide), Xixianmalin
IUPAC Name 3-(4-chlorophenyl)-3-(3,4-dimethoxyphenyl)-1-morpholin-4-ylprop-2-en-1-one
SMILES COc1ccc(cc1OC)C(=CC(=O)N2CCOCC2)c3ccc(Cl)cc3
Specifications
Quantity 1 g
InChI Formula InChI=1S/C21H22ClNO4/c1-25-19-8-5-16(13-20(19)26-2)18(15-3-6-17(22)7-4-15)14-21(24)23-9-11-27-12-10-23/h3-8,13-14H,9-12H2,1-2H3
Formula Weight 387.1237
Percent Purity >95%
Chemical Name or Material Dimethomorph
Grade HPLC