missing translation for 'onlineSavingsMsg'
Learn More

Dimenhydrinate, MP Biomedicals™

Product Code. p-4405478 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11451611

Brand: MP Biomedicals 0219018910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 523-87-5
Molecular Formula C24H28ClN5O3
Molecular Weight (g/mol) 469.97
InChI Key NFLLKCVHYJRNRH-UHFFFAOYSA-N
Synonym dimenhydrinate, dramamine, vomex a, chloranautine, diphenhydrinate, anautine, travelin, amosyt, aviomarin, gravol
PubChem CID 10660
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine;8-chloro-1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Cl.CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Color White
Quantity 10 g
Formula Weight 470
Physical Form Powder
Chemical Name or Material Dimenhydrinate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.