Learn More
Dimemorfan phosphate, MedChemExpress
Product Code. 16444657
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444657 50 mg 50mg
16438876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16444657 Supplier Medchem Express Supplier No. HYB2215
MedChemExpress Dimemorfan phosphate is a sigma 1 receptor agonist, used as a potent antitussive.
Dimemorfan phosphate is a sigma 1 receptor agonist, used as a potent antitussive.
Specifications
Chemical Name or Material Dimemorfan phosphate
CAS 36304-84-4
Color White
Purity Grade Notes Research
Molecular Formula C18H28NO4P
Quantity 50 mg
Solubility Information H2O : 10 mg/mL (28.30 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 1 mg/mL (2.83 mM; Need ultrasonic)
SMILES O=P(O)(O)O.CN1[C@](CC2=CC=C(C)C=C32)([H])[C@](CCCC4)([H])[C@]34CC1
Molecular Weight (g/mol) 353.39
Formula Weight 353.39
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation