missing translation for 'onlineSavingsMsg'
Learn More

Dimemorfan phosphate, MedChemExpress

Product Code. 16438876
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438876 100 mg 100mg
16444657 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438876 Supplier Medchem Express Supplier No. HYB2215

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dimemorfan phosphate is a sigma 1 receptor agonist, used as a potent antitussive.

Dimemorfan phosphate is a sigma 1 receptor agonist, used as a potent antitussive.

Specifications

Chemical Name or Material Dimemorfan phosphate
CAS 36304-84-4
Color White
Purity Grade Notes Research
Molecular Formula C18H28NO4P
Quantity 100 mg
Solubility Information H2O : 10 mg/mL (28.30 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 1 mg/mL (2.83 mM; Need ultrasonic)
SMILES O=P(O)(O)O.CN1[C@](CC2=CC=C(C)C=C32)([H])[C@](CCCC4)([H])[C@]34CC1
Molecular Weight (g/mol) 353.39
Formula Weight 353.39
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.