Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V6 Pumps
Product Code. 15288483
Change view
Click to view available options
Volume (Metric):
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
25 mL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
15288483 100 μL Each
15218483 50 μL Each
15238473 25 μL Each
15238493 250 μL Each
15238513 5 mL Each
15248503 1 mL Each
15278513 10 mL Each
15288503 2.5 mL Each
15288513 25 mL Each
15298493 500 μL Each
10 options
Product Code. 15288483
Brand: Trajan Scientific and Medical™ 005696
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for V6 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for V6 Pumps
Specifications
Description 60mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger
Length (Metric) 60 mm
Volume (Metric) 100 μL
For Use With (Equipment) V6 Pump