Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V6 Pumps
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Diluter/Dispenser Syringe for V6 Pumps
Specifications
Specifications
| Description | 60mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger |
| Volume (Metric) | 10 mL |
| For Use With (Equipment) | V6 Pump |