Learn More
Interscience™ DiluFlow Pro, Single Pump
Product Code. 17895070
Change view
Click to view available options
Accuracy:
>99%
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Accuracy Unit Size
17895070 >99% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17895070 Supplier Interscience™ Supplier No. 502105
  • ONLINE EXCLUSIVE PRICE
    81215.00 NOK
    79187.30 NOK / Pack of 1
    Save 2027.70 NOK
    2% Off
 Restricted Item
Estimated Shipment: 05-01-2027
The DiluFlow Pro automatically dilutes a solid sample in seconds, with the appropriate weight of diluent, up to a total weight of 5 kg. Its robotic arm enables easy work under a laminar flow cabinet.
  • With the press of only 2 keys, the DiluFlow Pro gravimetric dilutor provides an accurate dilution of all types of samples from 10 to 375 g (at 1/10th)
  • In compliance with ISO 7218 and ISO 6887
  • Robotic arm enables you to place the sample safely in the bag without risk of cross contamination
  • Widest possible opening for the bag and lowest loading threshold
  • With redesigned ergonomics, the ultra-low profile allows optimal comfort of use, especially under a laminar flow hood
  • In case of splashing, the drain tray is removable for rinsing and avoids tedious cleaning
  • GL45 (6.4 mm) per pump
Specifications
Accuracy >99%
Product Type Gravimetric Dilutor
Autoclavable Autoclavable
Certifications/Compliance ISO 7218, ISO 6887
Dilution Fast, Standard, Accurate
Volume (Metric) 100 to 5000 mL
For Use With (Application) Diluting Samples
Includes Drip Tray 400, 1 or 2 Pumps, Protection for Distribution Nozzle, Distribution Assembly, USB Cable, Supervision Software, Power Cord, BagOpen 400 for Dilutors, User Manual, Conformity Certificate/Warranty Card, 10 GeckoGrips, 1 Pk. of Adhesive Putty, Allen Wrench, Blender Bag Samples
Precision >99%
Speed 22.5 mL/sec.
Dimensions (L x W x H) 31.3 x 42.1 x 37.7 cm
Voltage 100/240 V
Wattage 60 W
Amperage 15 A