missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Diltiazem hydrochloride, 98%

Product Code. p-3778334 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10276161 1 g Glass bottle 1g
10464945 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10276161 Supplier Thermo Scientific Chemicals Supplier No. 329080010
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33286-22-5 | C22H26N2O4S·HCl | 450.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33286-22-5
Molecular Formula C22H26N2O4S·HCl
Molecular Weight (g/mol) 450.99
MDL Number MFCD00069252
InChI Key HDRXZJPWHTXQRI-BHDTVMLSSA-N
Synonym diltiazem hydrochloride, diltiazem hcl, herbesser, dilzene, lacerol, masdil, tildiem, tiazac, mono-tildiem, bi-tildiem
PubChem CID 62920
ChEBI CHEBI:645509
IUPAC Name [(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride
SMILES CC(=O)OC1C(SC2=CC=CC=C2N(C1=O)CCN(C)C)C3=CC=C(C=C3)OC.Cl

Specifications

Melting Point 210.0°C to 215.0°C
Color White
Ignition Residue 0.1% max.
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,3224
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,formic acid,methanol
Formula Weight 450.99
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Diltiazem hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Dilthiazem hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.