Learn More
Dilmapimod, MedChemExpress
Product Code. 16463938
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463938 10 mg 10mg
16446657 5 mg 5mg
16421049 1 mg 1mg
16400546 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16463938 Supplier Medchem Express Supplier No. HY10404
MedChemExpress Dilmapimod (SB-681323) is a potent p38 MAPK inhibitor that potentially suppresses inflammation in chronic obstructive pulmonary disease.
Dilmapimod (SB-681323) is a potent p38 MAPK inhibitor that potentially suppresses inflammation in chronic obstructive pulmonary disease.
Specifications
Chemical Name or Material Dilmapimod
CAS 444606-18-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H19F3N4O3
Quantity 10 mg
Synonym SB-681323 GW 681323
Solubility Information DMSO : 125 mg/mL (273.87 mM; Need ultrasonic)
SMILES O=C1C=CC2=C(C3=CC=C(F)C=C3C)N=C(NC(CO)CO)N=C2N1C4=C(F)C=CC=C4F
Molecular Weight (g/mol) 456.42
Formula Weight 456.42
Percent Purity 99.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation