Learn More
Diisobutyl Ketone, Sigma-Aldrich™
Specifications
Specifications
| CAS | 108-83-8 |
| Melting Point | -41°C |
| Density | 0.81 g/cm3 at 20°C |
| Boiling Point | 163°C to 173°C/1013 hPa |
| Molecular Formula | (CH3)2CHCH2COCH2CH(CH3)2 |
| MDL Number | MFCD00008940 |
| Quantity | 1 L |
| Synonym | Diisobutyl ketone, Di-iso-butyl ketone, 2,6-Dimethyl-4-heptanone, Isobutyl ketone, Diisobutyl ketone |
| Molecular Weight (g/mol) | 142.24 |
| Grade | Research |