Learn More
Dihexylamine, Sigma-Aldrich™
Specifications
Specifications
| CAS | 143-16-8 |
| Melting Point | 3°C |
| Density | 0.79 g/cm3 at 20°C |
| Boiling Point | 236°C/1013 mbar |
| Molecular Formula | CH3(CH2)5NH(CH2)5CH3 |
| MDL Number | MFCD00009521 |
| Quantity | 100 mL |
| Synonym | Dihexylamine, Dihexylamine |
| Molecular Weight (g/mol) | 185.35 |
| Grade | Research |