Learn More
Diglycidyl Resorcinol Ether (>85%), TRC
Product Code. 30377908
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377908 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30377908 Supplier Toronto Research Chemicals Supplier No. TRCD4463435ML
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Resorcinol diglycidyl ether
CAS 101-90-6
Molecular Formula C12 H14 O4
Quantity 5 mL
Synonym Oxirane, 2,2'-[1,3-phenylenebis(oxymethylene)]bis-, Benzene, m-bis(2,3-epoxypropoxy)- (6CI,7CI,8CI), 2,2'-[1,3-Phenylenebis(oxymethylene)]bis[oxirane], 1,3-Bis(2,3-epoxypropoxy)benzene, 1,3-Diglycidyloxybenzene, 1,3-Dihydroxybenzene diglycidyl ether, Diglycidyl resorcinol ether, NSC 76621, Resorcinol bis(2,3-epoxypropyl) ether, Resorcinol diglycidyl ether, Resorcinol glycidyl ether, m-Bis(2,3-epoxypropoxy)benzene, m-Bis(glycidyloxy)benzene
InChI Formula InChI=1S/C12H14O4/c1-2-9(13-5-11-7-15-11)4-10(3-1)14-6-12-8-16-12/h1-4,11-12H,5-8H2
SMILES C(Oc1cccc(OCC2CO2)c1)C3CO3
IUPAC Name 2-[[3-(oxiran-2-ylmethoxy)phenoxy]methyl]oxirane
Molecular Weight (g/mol) 222.24
Formula Weight 222.0892