Learn More
SI Analytics™ Digital Multi-Parameter Portable Meter
Description
- Parameters: pH, mV, O2 (saturation, concentration, partial pressure), Conductivity (spec. resistance, salinity, TDS), Temperature, Turbidity
- 1 universal measuring channel
- Protection class IP 67
- Two field-suitable waterproof USB interfaces for connecting memory sticks or selected printers and for data transfer to PC
Specifications
Specifications
| Data Management Details | Manual, time controlled data logger |
| Description | Mini USB-B interface |
| Display Type | Graphic, BW |
| Memory | Manual: 500 data sets; Automatic: 4500 data sets |
| Model | Multi 3510 IDS SET4 |
| Power Supply | 4 alkaline batteries, USB |