Learn More
Cole-Parmer™ Digi-Sense™ Detachable Thermocouple Probes
Product Code. 11820010 Shop All Cole-Parmer Products
Change view
Click to view available options
Depth (Metric):
17 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
11820010 17 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11820010 Supplier Cole-Parmer™ Supplier No. 0811740
  • ONLINE EXCLUSIVE PRICE
    436.00 NOK
    411.03 NOK / Pack of 1
    Save 24.97 NOK
    5% Off
View all promotions
Estimated Shipment: 26-01-2027
Attach these probes directly to your meter or probe handles. Digi-Sense™ Detachable Thermocouple Probes are ideal for measuring liquids, gases or semisolids.
Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue and type E-purple. Response Time: 2 seconds or greater.
Specifications
Temperature (Metric) -150°C to +649°C
Height (Metric) 90 mm
Depth (Metric) 17 mm