Learn More
Digi-Sense™ Precalibrated Pressure and Flow Meter
Product Code. 15850026
Change view
Click to view available options
Display Type:
LCD
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type Unit Size
15850026 LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15850026 Supplier Digi-Sense™ Supplier No. 2025013
  • ONLINE EXCLUSIVE PRICE
    4650.00 NOK
    4551.36 NOK / Pack of 1
    Save 98.64 NOK
    2% Off
Estimated Shipment: 29-12-2026
Includes
pitot tube, two rubber hoses for use with pitot tube, USB cable, software CD, ABS plastic carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
Easily determine the performance of your heating and cooling systems
  • Reliable measurements guaranteed
  • Precalibrated to NIST-traceable standards
  • Large, multiline backlit LCD
  • Save and recall 99 sample readings per parameter
  • Time clock on Max/Min/Avg provides time reference
Warranty
2 Years
Specifications
For Use With (Application) HVAC
Height (English) 2 in.
Height (Metric) 5 cm
Includes pitot tube, two rubber hoses for use with pitot tube, USB cable, software CD, ABS plastic carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.
Length (English) 8 in.
Length (Metric) 20 cm
Operating Temperature (English) 32°F to 122°F
Operating Temperature (Metric) 0°C to 50°C
Size (English) Fitting 1/4 in.
Size (Metric) Fitting 6.4 mm hose Barb
Width (English) 3 in.
Width (Metric) 7.5 cm
Accuracy ±0.3% Full-scale
Display Type LCD
Pressure Range 0 to 0.7252 psi
Product Type Digital Manometer