Learn More
Cole-Parmer™ Digi-Sense™ FEP-Insulated Type-K Probes with FEP-Coated Junction
Product Code. 11751349 Shop All Cole-Parmer Products
Change view
Click to view available options
Insulation:
FEP
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Insulation Unit Size
11751349 FEP Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11751349 Supplier Cole-Parmer™ Supplier No. 0846682
  • ONLINE EXCLUSIVE PRICE
    517.00 NOK
    488.05 NOK / Pack of 1
    Save 28.95 NOK
    5% Off
View all promotions
Estimated Shipment: 05-01-2027
Use these probes for applications requiring an isolated junction. Cole-Parmer™ Digi-Sense™ FEP-Insulated Type-K Probes with FEP-Coated Junction are ideal for long term immersion in semi caustic environments.
Due to the flexibility along with the sealed tip these probes can be used in harsh environments, submerged in liquid or great for general purpose everyday use. Type “K” is the most popular calibration thermocouple. It has the widest temperature range of all calibrations. Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue and type E-purple. Response Time: 2 seconds or greater.
  • Easily cleanable for multiple uses in different applications
  • Made from 24AWG cable for a fast response time along with the ability applied in hard to reach locations
  • Insulated wire probe single wire FEP with FEP over jacket
Specifications
Temperature (Metric) -150°C to +204°C
Insulation FEP