Learn More
SCHOTT AG Lighting and Imaging™ Diffuse Light Panel for Fiber Optic Flexible Light Guide
Product Code. 15783042 Shop All Schott Products
Change view
Click to view available options
Length (Metric):
78 mm
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15783042 78 mm 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15783042 Supplier SCHOTT AG Lighting and Imaging™ Supplier No. 500100
Estimated Shipment: 22-12-2026
Use the Diffuse Light Panel for Fiber Optic Flexible Light Guide for illumination of polished or high reflective surfaces. Additional flexible light guide required (155103 or 155104).
  • Diffuse and uniform illumination within a certain active area.
  • A white acrylic diffuser plate provides an optimum combination of uniformity and intensity.
  • Mounts directly to respective flexible light guide by means of thumb screw.
  • Housing made of rugged, black anodized aluminum.
  • For use with Cold light source KL1600LED, KL2500LED or KL1500HAL.
  • Dimensions: 137 x 58.5 x 23 mm Housing, 78 x 30 mm Active Area
Specifications
Type Diffuse Facelight
Dimensions (L x W x H) 137 x 58.5 x 23 mm Housing, 78 x 30 mm Active Area
For Use With (Application) Stereo Microscopy Illumination
For Use With (Equipment) KL1500 HAL, KL1600 LED, KL2500 LED Cold Light Sources
Includes 2 x M6 holes for articulated arm (158 345)
Length (English) 3.1 in.
Length (Metric) 78 mm
Width (English) 1.2 in.
Width (Metric) 30 mm