Learn More
BD Difco™ Disposable Inoculating Needles and Loops
Sterile, disposable inoculating needles do not require flaming
650.00 NOK - 825.00 NOK
Product Code | Brand | Color | Volume | Quantity | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume | Quantity | Product Type | Price | Quantity & Availability | ||
10492683
|
BD
220214 |
Light Green | 1 μL | 250/Pk. | Inoculating Loop |
825.00 NOK
Pack of 250 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10389822
|
BD
220216 |
Light Blue | 10 μL | 250/Pk. | Inoculating Loop |
650.00 NOK
Pack of 250 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
Description
- Made from a unique plastic resin for controlled flexibility
- Resist cutting the agar surface, yet provide sufficient firmness to pick stubborn colonies
- Free of lubricants, oils and electrostatic charges
- Color coded
Specifications
Plastic Resin | |
Sterile | |
Yes |