Learn More
BD Difco™ Differential Reinforced Clostridial Agar
Product Code. 11759062
Click to view available options
:
Clostridial Medium; Quantity: 500 g; Format: Dehydrated
Product Code. 11759062
Brand: BD 264120
BD Difco™ Differential Reinforced Clostridial Agar for cultivating and enumerating clostridia, other anaerobes, and other species of bacteria from foods and clinical specimens.
  • Dehydrated Culture Media
  • Formulation: Peptone 10 g/L, Beef extract 10 g/L, Yeast extract 3 g/L, Dextrose 5 g/L, Sodium Chloride 5 g/L, Soluble Starch 1 g/L, Cysteine HCl 0.5 g, Sodium Acetate 3 g, Agar 0.5 g/L
  • Certifications: CCAM, EP, JP, USP
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CCAM, EP, JP, USP
Form Dehydrated
Media Type Reinforced Clostridial Medium
Formulation Peptone 10 g/L, Beef extract 10 g/L, Yeast extract 3 g/L, Dextrose 5 g/L, Sodium Chloride 5 g/L, Soluble Starch 1 g/L, Cysteine HCl 0.5 g, Sodium Acetate 3 g, Agar 0.5 g/L
Packaging Type Bottle
Quantity 500 g