Learn More
BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium
Product Code. 11782834
Click to view available options
Quantity:
10,000 g
2000 g
500 g
Packaging Type:
Bottle
Pail
Media Type:
Fluid Thioglycollate Medium
Unit Size:
10kg
2kg
500g
Product Code. 11782834
Brand: BD 225650
For sterility testing of biologics and cultivation of anaerobes, aerobes and microaerophiles
  • Formulation: Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
Certifications
AOAC, BAM, BS12, COMPF, EP, USDA, USP
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance AOAC, BAM, BS12, COMPF, EP, USDA, USP
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Fluid Thioglycollate Medium
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
For Use With (Application) Used for sterility testing
Packaging Type Bottle
Quantity 500 g