Learn More
BD Difco™ Antibiotic Medium
Product Code. 11729092
Change view
Click to view available options
:
Antibiotic Medium; Quantity: 500 g; Format: Dehydrated
Product Code.
11729092 Antibiotic Medium; Quantity: 500 g; Format: Dehydrated
1 options
Product Code. 11729092
Brand: BD 266910
BD Difco™ Antibiotic Medium No. 12 is a microbiological assay of antibiotics.
  • Antibiotic assay media are used for determining antibiotic potency by the microbiological assay technique.
  • Antibiotic medium 12 is prepared with the grove and randall formula. It is used for the assay of amphotericin B.
  • Recommended for use in preparing test plates for the cylinder plate assay of nystatin and anisomycin.
  • Certifications: Antibiotic media are prepared according to the specifications of the USP, European Pharmacopeia, and AOAC International.
TRUSTED_SUSTAINABILITY
Specifications
Form Dehydrated
Quantity 500 g