missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriaminepentaacetic acid dianhydride, 95%

Product Code. p-7058432
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318186 1 g 1g
11328186 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318186 Supplier Thermo Scientific Alfa Aesar Supplier No. L15178.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23911-26-4 | C14H19N3O8 | 357.319 g/mol

Diethylenetriaminepentaacetic acid dianhydride is a reagent used for introducing a strong chelator. It is employed in the synthesis of macrocyclic ligands and lanthanide complexes that are useful as MRI enhancers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylenetriaminepentaacetic acid dianhydride is a reagent used for introducing a strong chelator. It is employed in the synthesis of macrocyclic ligands and lanthanide complexes that are useful as MRI enhancers.

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is-20°C. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23911-26-4
Molecular Formula C14H19N3O8
Molecular Weight (g/mol) 357.319
MDL Number MFCD00010697
InChI Key RAZLJUXJEOEYAM-UHFFFAOYSA-N
Synonym dtpa-anhydride, cyclic dtpa anhydride, 2-bis 2-2,6-dioxomorpholino ethyl amino acetic acid, unii-wnm9rlh5x9, wnm9rlh5x9, diethylenetriaminepentaacetic dianhydride, diethylenetriaminepentaacetic acid dianhydride, cdtpaa, glycine, n,n-bis 2-2,6-dioxo-4-morpholinyl ethyl, bis 2-2,6-dioxomorpholin-4-yl ethyl amino acetic acid
PubChem CID 100825
IUPAC Name 2-[bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino]acetic acid
SMILES C1C(=O)OC(=O)CN1CCN(CCN2CC(=O)OC(=O)C2)CC(=O)O

Specifications

Melting Point ∽190°C (decomposition)
Quantity 1 g
Beilstein 1230954
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 357.32
Percent Purity 95%
Chemical Name or Material Diethylenetriaminepentaacetic acid dianhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.