Learn More
Diethylene glycol divinyl ether, 98%, stab. 0.1% potassium hydroxide
Product Code. 11333196
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333196 50 g 50g
11323196 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11333196 Supplier Thermo Scientific Alfa Aesar Supplier No. L12501.18
 Restricted Item
In Stock
Add to basket
CAS: 764-99-8 | C8H14O3 | 158.197 g/mol
Diethylene glycol divinyl ether is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylene glycol divinyl ether is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 764-99-8
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00054610
InChI Key SAMJGBVVQUEMGC-UHFFFAOYSA-N
Synonym diethylene glycol divinyl ether, divinylcarbitol, dvedeg, dvedeg russian, ether, bis 2-vinyloxy ethyl, ethene, 1,1'-oxybis 2,1-ethanediyloxy bis, 3,6,9-trioxaundeca-1,10-diene, unii-abm90y2g95, divinyl ether diethylenglykolu, bis 2-vinyloxy ethyl ether
PubChem CID 12998
IUPAC Name 1-ethenoxy-2-(2-ethenoxyethoxy)ethane
SMILES C=COCCOCCOC=C
Specifications
Melting Point -21°C
Density 0.968
Boiling Point 198°C to 199°C
Flash Point 83°C (181°F)
Refractive Index 1.447
Quantity 50 g
Beilstein 1752316
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material Diethylene glycol divinyl ether, stabilized 0.1% potassium hydroxide