Learn More
Diethylammonium Chloride, Sigma-Aldrich™
Product Code. 15870931
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15870931 1 kg 1kg
15850931 5 g 5g
15860931 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15870931 Supplier Sigma Aldrich Supplier No. 8203931000
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 660-68-4
Specifications
Melting Point 231.8°C
Density 1.07 g/cm3 at 20.1°C
Molecular Formula (C2H5)2NH2Cl
MDL Number MFCD00012499
Quantity 1 kg
Synonym Diethylammonium chloride, Diethylamine hydrochloride, Diethylammonium chloride
Molecular Weight (g/mol) 109.6
Grade Research