missing translation for 'onlineSavingsMsg'
Learn More

Diethyl methylphosphonate, 97%, Thermo Scientific Chemicals

Catalog Number 11497836
Click to view available options
Quantity:
10 g
25 g
50 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
683-08-9
C5H13O3P
152.13
MFCD00009813
NYYLZXREFNYPKB-UHFFFAOYSA-N
diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h
12685
1-[ethoxy(methyl)phosphoryl]oxyethane
CCOP(C)(=O)OCC
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
683-08-9
C5H13O3P
152.13
MFCD00009813
NYYLZXREFNYPKB-UHFFFAOYSA-N
diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h
12685
1-[ethoxy(methyl)phosphoryl]oxyethane
CCOP(C)(=O)OCC

Diethyl methylphosphonate is used as a precursor for the synthesis of gem-difluroalkenyphosphonate via reaction with alpha-trifluromethylstyrene. It is also used in the synthesis of phosphonylated peptides, fluoroalkyl alpha- and beta-aminophosphonates, pyridone alkaloids with neuritogenic activity and lipophilic meropenem-derived prodrugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl methylphosphonate is used as a precursor for the synthesis of gem-difluroalkenyphosphonate via reaction with alpha-trifluromethylstyrene. It is also used in the synthesis of phosphonylated peptides, fluoroalkyl alpha- and beta-aminophosphonates, pyridone alkaloids with neuritogenic activity and lipophilic meropenem-derived prodrugs.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Density 1.04
Boiling Point 194°C
Flash Point 75°C (167°F)
Refractive Index 1.414
Quantity 10 g
Beilstein 1753416
Solubility Information Miscible with water.
Formula Weight 152.13
Percent Purity 97%
Chemical Name or Material Diethyl methylphosphonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'