missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, Supelco™

Product Code. 15896900
Change view
Click to view available options
Quantity:
1 L
25 L
Unit Size:
1 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896900 25 L 25 L
15893970 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15896900 Supplier Supelco Supplier No. 1009236025
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7

Specifications

Melting Point −116°C (lit.)
Density 0.706 g/mL at 25°C
Quantity 25 L
Molecular Formula (CH3CH2)2O
MDL Number MFCD00011646
Synonym Et2O, Ethoxyethane, Ethyl ether, Ether, Ether, Ethyl ether, Ether, Ethyl ether
Boiling Point 34.6°C
Molecular Weight (g/mol) 74.12
Grade Research
Chemical Name or Material Diethyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.