Learn More
Diethyl 3,4-pyridinedicarboxylate, 97%, Thermo Scientific™
Product Code. 10483074 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. Quantity Packaging unitSize
10483074 5 g Glass bottle 5g
1 options
Product Code. 10483074
Brand: Thermo Scientific Acros 399890050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 1678-52-0
Color Colorless to Yellow
Density 1.1450g/mL
Boiling Point 172°C (21.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C11H13NO4
MDL Number MFCD00012349
Quantity 5 g
Specific Gravity 1.145
Synonym diethyl 3,4-pyridinedicarboxylate, cinchomeronic ester, 3,4-pyridinedicarboxylic acid, diethyl ester, diethyl 3 4-pyridinedicarboxylate, ethyl 4-ethoxycarbonyl pyridine-3-carboxylate, 3,4-diethyl pyridine-3,4-dicarboxylate, pycb06, ksc532o9b, diethylpyridine-3,4-dicarboxylate
InChI Key SIOSRDUNOOIMCE-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(C=NC=C1)C(=O)OCC
IUPAC Name diethyl pyridine-3,4-dicarboxylate
Molecular Weight (g/mol) 223.23
PubChem CID 676160
Formula Weight 223.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl 3,4-pyridinedicarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl 3,4-pyridinedicarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.