Learn More
Diethyl 2-bromoethylphosphonate, 97%, Thermo Scientific Chemicals
Catalog Number p-7025592
Click to view available options
Quantity:
5 g
25 g
5324-30-1
C6H14BrO3P
245.05
MFCD00000239
PINITSMLVXAASM-UHFFFAOYSA-N
diethyl 2-bromoethyl phosphonate, diethyl 2-bromoethylphosphonate, phosphonic acid, 2-bromoethyl-, diethyl ester, 2-bromoethyl phosphonic acid diethyl ester, o,o-diethyl 2-bromoethyl phosphonate, diethyl bromoethylphosphonate, diethyl-2-bromoethyl-phosphonate, pinitsmlvxaasm-uhfffaoysa, diethyl-2-bromoethyl phosphonate
79218
1-bromo-2-diethoxyphosphorylethane
CCOP(=O)(CCBr)OCC
5324-30-1
C6H14BrO3P
245.05
MFCD00000239
PINITSMLVXAASM-UHFFFAOYSA-N
diethyl 2-bromoethyl phosphonate, diethyl 2-bromoethylphosphonate, phosphonic acid, 2-bromoethyl-, diethyl ester, 2-bromoethyl phosphonic acid diethyl ester, o,o-diethyl 2-bromoethyl phosphonate, diethyl bromoethylphosphonate, diethyl-2-bromoethyl-phosphonate, pinitsmlvxaasm-uhfffaoysa, diethyl-2-bromoethyl phosphonate
79218
1-bromo-2-diethoxyphosphorylethane
CCOP(=O)(CCBr)OCC
It is employed as a reactant involved in radical coupling, synthesis of organosoluble zirconium phosphonate nanocomposites, click-chemistry, asymmetric epoxidation of unfunctionalized olefins, Negishi alkyl-aryl cross-coupling, and in synthesis of hydrolytically stable phosphonic acids for use as dental adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a reactant involved in radical coupling, synthesis of organosoluble zirconium phosphonate nanocomposites, click-chemistry, asymmetric epoxidation of unfunctionalized olefins, Negishi alkyl-aryl cross-coupling, and in synthesis of hydrolytically stable phosphonic acids for use as dental adhesives.
Solubility
Soluble in water(20 g/L) (25°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Density 1.275
Boiling Point 98°C to 100°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4605
Quantity 5 g
Beilstein 1766077
Solubility Information Soluble in water(20g/L) (25°C).
Formula Weight 245.06
Percent Purity 97%
Chemical Name or Material Diethyl 2-bromoethylphosphonate