Learn More
Diethyl 2-Acetamidomalonate, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Diethyl Acetamidomalonate |
| CAS | 1068-90-2 |
| Molecular Formula | C9 H15 N O5 |
| Quantity | 50 g |
| Synonym | Propanedioic acid, 2-(acetylamino)-, 1,3-diethyl ester, Malonic acid, acetamido-, diethyl ester (6CI,7CI,8CI), Propanedioic acid, (acetylamino)-, diethyl ester (9CI), (Acetylamino)malonic acid diethyl ester, (Acetylamino)propanedioic acid diethyl ester, 2-(Acetylamino)propanedioic acid diethyl ester, 2-Acetylamino-malonic acid diethyl ester, Acetamidomalonic acid diethyl ester, Diethyl (N-acetylamino)malonate, Diethyl (acetylamino)malonate, Diethyl 2-(acetamido)propanedioate, Diethyl 2-(acetylamino)propanedioate, Diethyl 2-acetamidomalonate, Diethyl 2-acetylaminomalonate, Diethyl acetamidomalonate, Diethyl α-(acetylamino)malonate, NSC 7645, 2-Acetamidomalonic acid-diethyl ester |
| InChI Formula | InChI=1S/C9H15NO5/c1-4-14-8(12)7(10-6(3)11)9(13)15-5-2/h7H,4-5H2,1-3H3,(H,10,11) |
| SMILES | CCOC(=O)C(NC(=O)C)C(=O)OCC |
| IUPAC Name | diethyl 2-acetamidopropanedioate |
| Molecular Weight (g/mol) | 217.22 |
| Formula Weight | 217.095 |