Learn More
Didodecyl Phthalate, TRC
Product Code. 17900075
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17900075 1 g 1g
17929734 5 g 5g
17988664 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17900075 Supplier Toronto Research Chemicals Supplier No. TRCD4417651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2432-90-8
Molecular Formula C32 H54 O4
Molecular Weight (g/mol) 502.77
Synonym Phthalic acid, bis-dodecyl ester, 1,2-Benzenedicarboxylic acid, 1,2-didodecyl ester (ACI), 1,2-Benzenedicarboxylic acid, didodecyl ester (9CI), Phthalic acid, didodecyl ester (6CI, 7CI, 8CI), Bis-didecyl phthalate, Di-n-dodecyl phthalate, Didodecyl phthalate, Dilauryl phthalate, NSC 68022, Vinycizer 126
IUPAC Name didodecyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCCCCC
Specifications
Quantity 1 g
InChI Formula InChI=1S/C32H54O4/c1-3-5-7-9-11-13-15-17-19-23-27-35-31(33)29-25-21-22-26-30(29)32(34)36-28-24-20-18-16-14-12-10-8-6-4-2/h21-22,25-26H,3-20,23-24,27-28H2,1-2H3
Formula Weight 502.4022
Chemical Name or Material Phthalic acid, bis-dodecyl ester