Learn More
Thermo Scientific Cap AL Sealing Tube 2ml
Brand:  Thermo Scientific 54459
Cap AL Sealing Tube 2ml
Product Code. 10004643
  • ONLINE EXCLUSIVE PRICE
    7545.00 NOK
    7335.13 NOK / Each
    Save 209.87 NOK
    2% Off