Learn More
Dichloromethane-D2, Sigma-Aldrich™
Description
A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.
Specifications
Specifications
| Density | 1.36 g/cm3 at 20°C |
| Quantity | 10 x 0.75 mL |
| CAS | 1665-00-5 |
| Melting Point | -97°C |
| Boiling Point | 39°C/1013 hPa |
| MDL Number | MFCD00000882 |
| Synonym | Dichloromethane-D2, Dideuteromethylene chloride, Methylene chloride-d2, Dideuterodichloromethane, DCM deuterated, Dichloromethane-D2 |
| Molecular Weight (g/mol) | 86.94 |
| Grade | Research |
| Chemical Name or Material | Dichloromethane-D2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.