Learn More
Dichlorodimethylsilane, Sigma-Aldrich™
Product Code. 15854131
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15854131 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15854131 Supplier Sigma Aldrich Supplier No. 8034520250
 Restricted Item
Estimated Shipment: 02-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 70°C/1013 hPa
Melting Point -76°C
CAS 75-78-5
Quantity 250 mL
Molecular Formula (CH3)2SiCl2
MDL Number MFCD00000491
Synonym Dichlorodimethylsilane, Dimethyldichlorosilane, Dichlorodimethylsilane
Molecular Weight (g/mol) 129.06
Grade Research
Density 1.07 g/cm3 at 25°C
Chemical Name or Material Dichlorodimethylsilane