Learn More
Dichloroacetic Acid, Sigma-Aldrich™
Specifications
Specifications
| CAS | 79-43-6 |
| Melting Point | -4°C |
| Density | 1.57 g/cm3 at 20°C |
| Boiling Point | 192°C/1013 hPa (Decomposes) |
| MDL Number | MFCD00004223 |
| Quantity | 5 mL |
| Synonym | Dichloroacetic acid, Dichloroacetic acid |
| Molecular Weight (g/mol) | 128.94 |
| Grade | Research |