Learn More
Dichloroacetic Acid-d2, CDN
Product Code. 30328422
Click to view available options
Quantity:
1 g
Unit Size:
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30328422
Brand: CDN CDND10951G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dichloroacetic Acid-d2
CAS 63403-57-6
Molecular Formula C2 D2 Cl2 O2
Quantity 1 g
Synonym Acetic-d acid-d, dichloro- (9 CI), 2,2-Dichloroacetic-2-d acid-d, Dichloroacetic acid-d2, Dichloroacetic Acid-D2
InChI Formula InChI=1 S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/i1D/hD
SMILES [2 H]OC(=O)C([2 H])(Cl)Cl
IUPAC Name deuterio 2,2-dichloro-2-deuterioacetate
Molecular Weight (g/mol) 130.9544
Formula Weight 129.9557 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity