Learn More
JT Baker Dichloromethane UltRA RESI-ANALYZED 1L
Brand:  JT Baker 9264.1000
Dichloromethane UltRA RESI-ANALYZED 1L
Product Code. 10140421
  • ONLINE EXCLUSIVE PRICE
    7160.00 NOK
    2405.63 NOK / 1 L
    Save 4754.37 NOK
    66% Off
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P281-Use personal protective equipment as required.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.