Learn More
Dibutyltin Dilaurate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 77-58-7 |
| Melting Point | 25°C to 27°C |
| Density | 1.05 g/cm3 at 20°C |
| Boiling Point | >250°C/1013 hPa |
| MDL Number | MFCD00008963 |
| Quantity | 250 mL |
| Synonym | Dibutyltin dilaurate, Dibutylbis(lauroyloxy)stannane, Dibutyl bis(lauroyloxy)tin, Dibutyltin dilaurate |
| Molecular Weight (g/mol) | 631.56 |
| Grade | Research |
| Chemical Name or Material | Dibutyltin Dilaurate |