missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin Dichloride, Sigma-Aldrich™

Product Code. 15834151
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15834151 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15834151 Supplier Sigma Aldrich Supplier No. 8041840250
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 683-18-1
Melting Point 37°C to 38°C
Density 1.4 g/cm3 at 20°C
Boiling Point 148°C/16 hPa
Molecular Formula (CH3CH2CH2CH2)2SnCl2
MDL Number MFCD00000518
Quantity 250 g
Synonym Dibutyltin dichloride, Dibutyldichlorostannane, Dibutyldichlorotin, Dibutyltin dichloride
Molecular Weight (g/mol) 303.84
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.